4-Sulfamoylbenzoic acid ≥95%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 4-Sulphamoylbenzoic acid

S1753-25GM S1753-100GM S1753-500GM
10860-308EA 50.9 USD
10860-308 10860-310 10860-312
4-Sulfamoylbenzoic acid ≥95%
4-Sulphamoylbenzoic acid

GHS hazard : Exclamation mark

Formula: C₇H₇NO₄S
MW: 201.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007938
CAS Number: 138-41-0

Order Now

Specification Test Results

Purity (Nonaqueous Titration) 95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR