4,4'-(Hexafluoroisopropylidene)diphthalic anhydride ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 5,5'-(Perfluoropropane-2,2-diyl)bis(isobenzofuran-1,3-dione), 2,2-Bis(1,3-dihydro-1,3-dioxobenzo[c]furan-5-yl)hexafluoropropane

H1784-5GM H1784-25GM
10841-852EA 99.05 USD
10841-852 10841-854
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride ≥98%
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride

Storage temperature - MOISTURE SENSITIVE: Keep tightly closed.


GHS hazard : Corrosion UN3261


Precaución: Storage temperature - MOISTURE SENSITIVE: Keep tightly closed.

Formula: C₈H₄O₃
Storage Temperature: Ambient
MDL Number: MFCD00039143
CAS Number: 1107-00-2
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Purity (GC) 98 %
Purity (Morpholine Method) 98 %
Melting Point 243.0°-249.0°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR