2-Octyl-1-dodecanol

Supplier: Spectrum Chemical Mfg. Corp.

O2014-500ML O2014-100ML O2014-25ML
700002-806EA 630.9 USD
700002-806 700002-802 700002-804
2-Octyl-1-dodecanol
2-Octyl-1-dodecanol

2-Octyl-1-dodecanol is widely used in cosmetics and pharmaceutical applications as an emulsifying and opacifying agent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₀H₄₂O
Density: 0.838
Storage Temperature: Ambient
MDL Number: MFCD01310428
CAS Number: 5333-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR