Naphthalene-2-sulfonic acid hydrate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-Naphthalenesulphonic acid hydrate

N2337-100GM N2337-25GM N2337-500GM
10841-722EA 182.55 USD
10841-722 10841-724 10841-726
Naphthalene-2-sulfonic acid hydrate
Naphthalene-2-sulfonic acid hydrate

GHS hazard : Corrosion UN2583

Formula: C₁₀H₈O₃S·xH₂O
MW: 208.231 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149271
CAS Number: 76530-12-6
UN: 2583
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR