Sodium-2-naphthalenesulfonate ≥90%, powder
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
2-Naphthalenesulphonic acid Sodium Salt, Sodium 2-naphthalenesulphonate
N2022-100GM
N2022-500GM
N2022-25GM
700002-514EA
309.3
USD
700002-514
700002-518
700002-516
Sodium-2-naphthalenesulfonate ≥90%, powder
Sodium-2-naphthalenesulfonate
2-Naphthalenesulfonic Acid Sodium Salt, Powder is an ion-pair reagent that is used as an additive in the mobile phase of HPLC to enhance the separation of ionic and highly polar substances.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₀H₈O₃S MW: 230.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064186 CAS Number: 532-02-5 |
Specification Test Results
Assay | 90 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...