2-Methyl-4-isothiazolin-3-one hydrochloride

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Methylisothiazolinone hydrochloride, MIT

M2105-1GM M2105-5GM
700008-146EA 500.65 USD
700008-146 700008-148
2-Methyl-4-isothiazolin-3-one hydrochloride
2-Methyl-4-isothiazolin-3-one hydrochloride

2-Methyl-4-isothiazolin-3-one Hydrochloride is used for its antibacterial properties.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄H₆ClNOS
MW: 151.608 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06804636
CAS Number: 26172-54-3
UN: 1759
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR