Iodoacetamide ≥98%, crystals
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
2-Iodoacetamide
I1038-100GM
I1038-25GM
I1038-5GM
10841-648EA
991.85
USD
10841-648
10841-718
10841-720
Iodoacetamide ≥98%, crystals
Iodoacetamide
2-Iodoacetamide, Crystal, an alkylating agent, is used for peptide mapping purposes.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
ICH₂CONH₂ MW: 184.96 g/mol Boiling Pt: 266 Storage Temperature: Refrigerator |
MDL Number:
MFCD00008028 CAS Number: 144-48-9 UN: 2811 ADR: 6.1 III |
Specification Test Results
Assay | 98 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...