2-Butyne-1,4-diol ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1,4-Dihydroxy-2-butyne, But-2-yne-1,4-diol

B2232-100GM B2232-500GM
10841-644EA 92.9 USD
10841-644 10841-646
2-Butyne-1,4-diol ≥99%
2-Butyne-1,4-diol

2-Butyne-1,4-diol is a colorless, hygroscopic solid that is soluble in water and polar organic solvents. It is primarily used as a synthetic organic intermediate used in the manufacture of certain herbicides, textile additives, corrosion inhibitors, plasticizers, synthetic resins, and polyurethanes. It is the major raw material of vitamin B6 and is also used for brightening, preserving, and inhibiting nickel plating


GHS hazard : Skull and crossbones;Exclamation mark;Health hazard
UN2716

Formula: C₄H₆O₂
MW: 86.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002915
CAS Number: 110-65-6
UN: 2716
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR