2,5-Dihydroxyterephthalic acid ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP

D2940-5GM D2940-25GM
10860-396EA 201.5 USD
10860-396 10860-398
2,5-Dihydroxyterephthalic acid ≥98%
2,5-Dihydroxyterephthalic acid

GHS hazard : Exclamation mark

Formula: C₈H₆O₆
MW: 198.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00132933
CAS Number: 610-92-4

Order Now

Specification Test Results

Purity (GC) 98 %
Purity (Neutralization Titration) 98 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR