2,4-Dichlorobenzyl alcohol, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (2,4-Dichlorophenyl)methanol

D2856-25GM D2856-100GM
10860-390EA 198.8 USD
10860-390 10860-392
2,4-Dichlorobenzyl alcohol, Reagent Grade
2,4-Dichlorobenzyl alcohol
Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004606
CAS Number: 1777-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR