2,3,6,7,10,11-Hexahydroxytriphenylene hydrate ≥95.0%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Triphenylene-2,3,6,7,10,11-hexaol hydrate

H1786-1GM H1786-5GM
10860-386EA 181.77 USD
10860-386 10860-388
2,3,6,7,10,11-Hexahydroxytriphenylene hydrate ≥95.0%
2,3,6,7,10,11-Hexahydroxytriphenylene hydrate

GHS hazard : Exclamation mark

Formula: C₁₈H₁₂O₆·xH₂O
MW: 324.29 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD01321170
CAS Number: 4877-80-9

Order Now

Specification Test Results

Purity (HPLC; Area %) 95.0 %
Water max.8.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR