Monomyristin

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1-Monomyristoyl Glycerol, Glycerol alpha-Monomyristate, 2,3-Dihydroxypropyl tetradecanoate

M1612-100GM M1612-500GM M1612-25GM
700005-236EA 314.75 USD
700005-236 700005-240 700005-238
Monomyristin
Monomyristin

1-Monomyristin is used as an emollient and emulsifying agent in personal care products, including cosmetics, shampoos, perfumes, soaps and lotions.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₇H₃₄O₄
MW: 302.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046760
CAS Number: 589-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR