2-Methoxy-1-methylethyl acetate (PGMEA) ≥97%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Propylene glycol 1-methyl ether 2-acetat, 2-(1-Methoxy)propyl acetate, 1-Methoxy-2-propyl acetate, Propylene glycol methyl ether acetat, 1,2-Propanediol monomethyl ether acetate, DOWANOL PMA

M2634-20LT M2634-4LT M2634-1LT
10841-620EA 820.85 USD
10841-620 10841-622 10841-636
2-Methoxy-1-methylethyl acetate (PGMEA) ≥97%
2-Methoxy-1-methylethyl acetate (PGMEA)

1-Methoxy-2-propanol Acetate is used as solvent in the semiconductor industry and in inks, coatings and cleaners.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₁₂O₃
Density: 0.968
Storage Temperature: Ambient
MDL Number: MFCD00038500
CAS Number: 108-65-6
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Assay 97 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR