1,4-Phenylenediacetic acid ≥98%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
2,2'-(1,4-Phenylene)diacetic acid
P2035-5GM
P2035-25GM
10860-372EA
111.25
USD
10860-372
10860-374
1,4-Phenylenediacetic acid ≥98%
1,4-Phenylenediacetic acid
GHS hazard : Exclamation mark
Formula:
C₁₀H₁₀O₄ MW: 194.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004354 CAS Number: 7325-46-4 |
Specification Test Results
Purity (HPLC) | 98 % |
Purity (Nonaqueous Titration) | 98 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...