1,3-Dioxolane ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

D2371-20LT D2371-4LT
10841-546EA 1019.35 USD
10841-546 10841-548
1,3-Dioxolane ≥99%
1,3-Dioxolane

1,3-Dioxolane finds great utility in many formulation as a powerful aprotic solvent with low toxicity and low boiling point (76 °C) and an inhibitor in 1,1,1-trichloroethane. Similar to most ethylene glycol-based ethers, 1,3-dioxolane can dissolve many inorganic salts. The product finds great utility in metal working and electroplating as brightening additive (Ni & Cu) and increasing the effectiveness of mineral etching solutions


GHS hazard : Flame;Exclamation mark
UN1166

Formula: C₃H₆O₂
Density: 1.064
Storage Temperature: Ambient
MDL Number: MFCD00003207
CAS Number: 646-06-0
UN: 1166
ADR: 3,II

Order Now

Specification Test Results

Assay 99 % min.
Color (APHA) max.10
Acidity (As CH₃COOH) max. 100 ppm
Peroxide max. 5 ppm
Water max. 150 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR