1,3,5-Tris(4-carboxyphenyl)benzene ≥99.0%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
5'-(4-Carboxyphenyl)-[1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid
T1642-1GM
T1642-5GM
10860-368EA
229.3
USD
10860-368
10860-370
1,3,5-Tris(4-carboxyphenyl)benzene ≥99.0%
1,3,5-Tris(4-carboxyphenyl)benzene
GHS hazard : Exclamation mark
Formula:
C₂₇H₁₈O₆ MW: 438.44 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10000888 CAS Number: 50446-44-1 |
Specification Test Results
Assay (HPLC) | 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...