(+/-)-Usnic acid ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Usnic acid, 2,6-Diacetyl-7,9-dihydroxy-8,9b-dimethyldibenzo[b,d]furan-1,3(2H,9bH)-dione

U1009-5GM U1009-25GM
700016-054EA 139.15 USD
700016-054 700016-052
(+/-)-Usnic acid ≥98%
(+/-)-Usnic acid

(+/-)-Usnic Acid, also known as usneine, is a dibenzofuran derivative used in perfumery, cosmetics and medicine as a preservative.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₈H₁₆O₇
MW: 344.3155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065294
CAS Number: 125-46-2

Order Now

Specification Test Results

Assay 98 % min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR