(±)-Camphorquinone ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dione, DL-Camphoroquinone

C2992-5GM C2992-25GM C2992-100GM
10860-356EA 82.15 USD
10860-356 10860-358 10860-360
(±)-Camphorquinone ≥98%
(±)-Camphorquinone
Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064160
CAS Number: 10373-78-1

Order Now

Specification Test Results

Purity (GC) 98 %
Melting Point 199.0°-203.0°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR