D-(+)-Menthol ≥99.0%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
(1S)-(+)-Menthol, (1R,2S,4S)-(+)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, (1S,2R,5S)-(+)-Menthol, D(+)-Menthyl alcoholol
M1768-100GM
M1768-25GM
10860-512EA
297.1
USD
10860-512
10860-514
D-(+)-Menthol ≥99.0%
D-(+)-Menthol
GHS hazard : Exclamation mark
Formula:
C₁₀H₂₀O MW: 156.27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00062983 CAS Number: 15356-60-2 |
Specification Test Results
Purity (GC) | 99.0 % min. |
Melting Point | 41.0° - 45.0°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...