Poloxamer 124 (Pluronic® L44)

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Pluronic® L44, PEG-PPG-PEG, Polyethylene-Polypropylene Glycol

P1168-20LT P1168-500MLG P1168-200LTB P1168-2.5LTG
700012-002EA 1327.1 USD
700012-002 700012-004 700012-000 700011-998
Poloxamer 124 (Pluronic® L44)
Poloxamer 124 (Pluronic® L44)

Poloxamer 124 has an amphiphilic structure (both hydrophilic and lipophilic) and is a useful surfactant for the formulation of cosmetics and pharmaceuticals because it increases miscibility. Poloxamer 124 has also been used to control thickness (viscosity) of personal care products and dish soaps.


Poloxamer 124 has also been used to control thickness (viscosity) of personal care products and dish soaps.

Formula: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
Storage Temperature: Ambient
MDL Number: MFCD00082049
CAS Number: 9003-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR