RiboGuard™ RNase Inhibitor
Supplier: Biosearch Technologies
RG90910K RG90925
75935-554EA 614.07 USD
75935-554 75935-556
RiboGuard™ RNase Inhibitor
Proteins and Peptides
Protect your precious RNA with this high quality, effective RNase inhibitor.
  • Valuable: Protects your RNA by inhibiting RNase A, RNase B and RNase C.
  • Compatible: Add to critical cDNA synthesis, RT-PCR, and in vitro transcription and translation reactions without inhibiting enzyme activity.
RiboGuard™ RNase Inhibitor is the best defense against common RNases, including RNase A, RNase B, and RNase C. This recombinant RNase inhibitor protein provides reliable protection of valuable RNA samples by binding strongly to RNases at a 1:1 ratio. Advanced purification protocols and extensive quality control tests ensure RiboGuard RNase Inhibitor is free of unwanted contaminants that can plague other commercially available RNase inhibitor preparations.
Información de suministro: Contains RiboGuard™ RNase Inhibitor at a concentration of 40 U/µL.
Precaución: For research use only. Not for human or diagnostic use.