Cefoxitin sodium

Supplier: CHEM IMPEX INTL INC MS

Synonyms: Mefoxin, Cefoxitin sodium salt, Sodium (6R,7S)-3-((carbamoyloxy)methyl)-7-methoxy-8-oxo-7-(2-(thiophen-2-yl)acetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

01490-1G 01490-5G 01490-1G 01490-5G
102616-182EA 27.05 USD
102616-182 102616-184 CH01490-1G CH01490-5G
Cefoxitin sodium
Cefoxitin sodium

Synonyms: Mefoxin

Formula: C₁₆H₁₆N₃NaO₇S₂
Storage Temperature: Refrigerator
MDL Number: MFCD00079042
CAS Number: 33564-30-6

Order Now

Specification Test Results

Appearance White to off-white powder
Crystallinity Meets requirements (695)
Heavy Metals ≤0.002%
Water Content (Karl Fisher) ≤1%
pH 4.2 - 7.0 (791)
Optical Rotation +206° to +214°
Residual Solvents (Acetone) ≤0.7%
Residual Solvents (Methanol) ≤0.1%
Bacterial Endotoxins ≤0.13 Units/mg
Particle Size (≥10 µm) ≤6000 pc/g
Particle Size (≥25 µm) ≤600 pc/g
Potency (Anhydrous Basis) ≥940 µg/mg (acetone and methanol free)
Purity by HPLC Conforms (with reference)
Purity by UV Conforms (197U)
Storage Temperature 0 - 8 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR