Butyl acetate ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate
270687-1L
270687-6X1L
BJ270687-1LEA
266
USD
BJ270687-1L
BJ270687-6X1L
Butyl acetate ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Butyl acetate
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities
Formula:
CH₃COO(CH₂)₃CH₃ MW: 116.16 g/mol Boiling Pt: 126.2 °C (1.013 hPa) Melting Pt: -78 °C Density: 0.8825 g/cm3 Flash Pt: 10.4 %(V) Storage Temperature: Ambient |
MDL Number:
MFCD00009445 CAS Number: 123-86-4 EINECS: 204-658-1 UN: 1123 ADR: 3,III |
Specification Test Results
Identity (IR) | complying |
Assay (GC) | Min. 99.65 % |
Non-volatile matter | Max. 0.0005 % |
Water (Karl Fischer) | Max. 0.05 % |
Absorbance at 400 nm | Max. 0.01 |
Absorbance at 320 nm | Max. 0.01 |
Absorbance at 300 nm | Max. 0.02 |
Absorbance at 275 nm | Max. 0.04 |
Absorbance at 260 nm | Max. 0.20 |
Absobance at 254 nm | Max. 1.0 |
Explosion Limit | 7.6 % |
Autoignition Temperature | 790 °F |
Vapor Pressure | 15 mmHg ( 25 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...