2-Methylbutane 99,5, CHROMASOLV™ HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: iso-Pentane

270342-1L 270342-2.5L
BJ270342-1LEA 297.52 USD
BJ270342-1L BJ270342-2.5L
2-Methylbutane 99,5, CHROMASOLV™ HPLC, Riedel-de Haën™
2-Methylbutane

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formula: CH₃CH₂CH(CH₃)₂
MW: 72.15 g/mol
Boiling Pt: 28
Melting Pt: -160
Density: 0.62 g/cm³ (20 °C)
Flash Pt: <–35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009338
CAS Number: 78-78-4
EINECS: 201-142-8
UN: 1265
ADR: 3 I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=498904a9f8b2618e19d2343aa74e247ec7121f00&vid=c7ed2044255d0d72af23a3fafec4ef6066d25097&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.50 %
Non-volatile matter Max. 0.0003 %
Water (Karl Fischer) Max. 0.02 %
Absorbance at 400 nm Max. 0.01
Absorbance at 240 nm Max. 0.01
Absorbance at 220 nm Max. 0.07
Absorbance at 210 nm Max. 0.30
Absorbance at 192 nm Max. 1.0
Vapor Density 2.6 (vs air)
Autoignition Temperature 788 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR