Cobalt(II) chloride hexahydrate 98% ACS, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
255599-100G
255599-500G
255599-5G
BJ255599-100GEA
107.46
USD
BJ255599-100G
BJ255599-500G
BJ255599-5G
Cobalt(II) chloride hexahydrate 98% ACS, Fluka™
Cobalt(II) chloride hexahydrate
Formula:
CoCl₂·6H₂O MW: 237.93 g/mol Boiling Pt: Dec. (on heating) Melting Pt: 86 °C Density: 1.9 g/cm3 Storage Temperature: Room Temperature |
MDL Number:
MFCD00149652 CAS Number: 7791-13-1 EINECS: 231-589-4 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 98.0 - 102.0 % |
Water insoluble matter | Max. 0.01 % |
Calcium (Ca) | Max. 0.005 % |
Copper (Cu) | Max. 0.002 % |
Iron (Fe) | Max. 0.005 % |
Potassium (K) | Max. 0.01 % |
Magnesium (Mg) | Max. 0.005 % |
Sodium (Na) | Max. 0.05 % |
Nickel (Ni) | Max. 0.1 % |
Zinc (Zn) | Max. 0.03 % |
Nitrate (NO₃) | Max. 0.01 % |
Sulfate (SO₄) | Max. 0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...