tert-Butyl methyl ether ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
MTBE, 2-Methoxy-2-methylpropane, Methyl-t-Butyl Ether
34875-100ML
34875-2.5L
34875-4X2.5L
34875-6X1L
34875-4X4L
BJ34875-100MLEA
91.54
USD
BJ34875-100ML
BJ34875-1L
BJ34875-2.5L
BJ34875-4X2.5L
BJ34875-6X1L
BJ34875-4X4L
tert-Butyl methyl ether ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
tert-Butyl methyl ether
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Beilstein Number: 1730942
Formula:
(CH₃)₃COCH₃ MW: 88.15 g/mol Boiling Pt: 55.3 °C (1.013 hPa) Melting Pt: -109 °C Density: 0.74 g/cm3 Flash Pt: 8.5 %(V) Storage Temperature: Ambient |
MDL Number:
MFCD00008812 CAS Number: 1634-04-4 EINECS: 216-653-1 UN: 2398 ADR: 3,II |
Specification Test Results
Assay (GC) | Min. 99.8 % |
IR spectrum | complying |
Non-volatile matter | Max. 0.0005 % |
Water (Karl Fischer) | Max. 0.01 % |
Free acid (as CH₃COOH) | Max. 0.002 % |
Peroxides (as H₂O₂) | Max. 0.0005 % |
Absorbance at 400 nm | Max. 0.005 |
Absorbance at 300 nm | Max. 0.005 |
Absorbance at 260 nm | Max. 0.05 |
Absorbance at 250 nm | Max. 0.10 |
Absorbance at 220 nm | Max. 0.40 |
Absorbance at 210 nm | Max. 1.0 |
APHA | Max. 10 |
Vapor Density | 3.1 (vs air) |
Refractive Index | n20/D 1.369 (lit.) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...