Toluene ≥99.7% (by GC), CHROMASOLV™ for pesticide residue analysis acc. to FDA, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Methylbenzene, Phenylmethane, FM approval

34494-1L 34494-4X2.5L
BJ34494-1LEA 72 USD
BJ34494-1L BJ34494-2.5L
Toluene ≥99.7% (by GC), CHROMASOLV™ for pesticide residue analysis acc. to FDA, Riedel-de Haën™
Toluene
Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6
Melting Pt: -95
Density: 0.867 g/cm³ (20 °C) (lit.)
Flash Pt: 7.1 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.7 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.02 %
Suitability for residue analysis Max. 5 ng/l
Refractive Index n20/D 1.496 (lit.)
Explosion Limit 7 %
Autoignition Temperature 997 °F
Vapor Pressure 26 mmHg ( 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR