Perdrogen® solution ≥30% (w/w) stabilized, Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
31642-5L
31642-6X1L
BJ31642-1LEA
253.11
USD
BJ31642-1L
BJ31642-500ML
BJ31642-5L
BJ31642-6X1L
Perdrogen® solution ≥30% (w/w) stabilized, Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™
Hydrogen peroxide
Honeywell ACS solvents for general laboratory use are:
- A value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- Variations and decrease issues traceable to solvent contamination
- All solvents are tested tmeet ACS general use specifications
Formula:
H₂O₂ Boiling Pt: Decomposes on heating Melting Pt: -26 °C Density: 1.11 g/cm3 Storage Temperature: Ambient |
MDL Number:
MFCD00011333 CAS Number: 7722-84-1 UN: 2014 ADR: 5.1,II |
Specification Test Results
Assay | Min. 30 % |
Non-volatile matter | Max. 0.005 % |
Residue on ignition | Max. 0.002 % |
Free acid (as H₂SO₄) | Max. 0.005 % |
Acidity | complying |
Aluminium (Al) | Max. 0.00005 % |
Arsenic (As) | Max. 0.000001 % |
Barium (Ba) | Max. 0.000005 % |
Beryllium (Be) | Max. 0.000001 % |
Bismuth (Bi) | Max. 0.00001 % |
Calcium (Ca) | Max. 0.00002 % |
Cadmium (Cd) | Max. 0.000001 % |
Cobalt (Co) | Max. 0.000001 % |
Chromium (Cr) | Max. 0.000002 % |
Copper (Cu) | Max. 0.000001 % |
Iron (Fe) | Max. 0.000005 % |
Germanium (Ge) | Max. 0.000005 % |
Potassium (K) | Max. 0.00001 % |
Lithium (Li) | Max. 0.000001 % |
Magnesium (Mg) | Max. 0.000005 % |
Manganese (Mn) | Max. 0.000001 % |
Molybdenum (Mo) | Max. 0.000002 % |
Sodium (Na) | Max. 0.00001 % |
Nickel (Ni) | Max. 0.000002 % |
Lead (Pb) | Max. 0.000001 % |
Strontium (Sr) | Max. 0.000001 % |
Titanium (Ti) | Max. 0.00001 % |
Thallium (Tl) | Max. 0.000005 % |
Vanadium (V) | Max. 0.000001 % |
Zinc (Zn) | Max. 0.000005 % |
Zirconium (Zr) | Max. 0.00001 % |
Chloride (Cl) | Max. 0.00005 % |
Phosphate (PO₄) | Max. 0.0001 % |
Sulfate (SO₄) | Max. 0.0001 % |
Total N | Max. 0.0002 % |
Organic stabilisators | Max. 0.05 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...