Ammonia solution 25 - 30% (NH₃ basis) in water, Puriss., meets analytical specification of Ph.Eur., Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
05003-1L
05003-2.5L
05003-25KG
BJ05003-1LEA
75.9
USD
BJ05003-1L
BJ05003-2.5L
BJ05003-25KG
BJ05003-5L
BJ05003-4X2.5L
BJ05003-4X5L
BJ05003-6X1L
Ammonia solution 25 - 30% (NH₃ basis) in water, Puriss., meets analytical specification of Ph.Eur., Fluka™
Ammonia solution
Formula:
NH₃ Boiling Pt: 37.7 °C (1.013 hPa) Melting Pt: -92 °C Density: 0.90 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00066650 CAS Number: 1336-21-6 UN: 2672 ADR: 8,III |
Specification Test Results
Assay | 25.0 - 30.0 % |
Density (D 20/20) | 0.892 - 0.910 |
Non-volatile matter | Max. 0.0018 % |
Iron (Fe) | Max. 0.25 ppm |
Heavy metals (as Pb) | Max. 1 ppm |
Carbonate (CO₃) | Max. 60 ppm |
Chloride (Cl) | Max. 1 ppm |
Sulfate (SO₄) | Max. 5 ppm |
Readily oxidizable substances | complying |
Pyridine, related substances (as Pyridine) | Max. 2 ppm |
Residual solvents | complying |
Appearance of the solution | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...