Aluminium oxide activated neutral Brockmann Grade I ~150 mesh, Fluka™
Supplier: Honeywell Research Chemicals
199974-1KG
199974-5KG
BJ199974-100GEA
64.38
USD
BJ199974-100G
BJ199974-1KG
BJ199974-5KG
Aluminium oxide activated neutral Brockmann Grade I ~150 mesh, Fluka™
Aluminium oxide activated neutral
Formula:
Al₂O₃ MW: 101.96 g/mol Boiling Pt: 2.980 °C (1.013 hPa) Melting Pt: 2.072 °C Density: 3.960 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 |
Specification Test Results
Brockmann Number (Activity) I (val.ref) | complying |
pH (10 % susp., 20°C) | 6.5 - 7.5 |
Spez. surface (BET) | 135 - 165 m²/g |
Fe (as Fe₂O₃) | Max. 0.03 % |
Na (as Na₂O) | Max. 0.4 % |
Silicon (as SiO₂) | Max. 0.03 % |
Chloride (Cl) | Max. 0.2 % |
Grain size 0.04-0.30 mm | Min. 95 % |
Pore Size | 58 +� pore size |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...