Lullaby™ Stem Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
LS20500
LS21000
103258-474EA
231.03
USD
103258-474
103258-476
Lullaby™ Stem Transfection Reagent, OZ Biosciences
Transfection Reagents
Lullaby Stem siRNA Transfection Reagent is ideal for gene silencing in Stem Cells.
- High gene silencing efficiency of multipotent and embryonic stem cells
- Minimized toxicity due to reagent biodegradability and low siRNA/miRNA amount required
- Reliable and reproducible gene knockdown results
- Serum & Antibiotics compatible
Lullaby Stem enables siRNA and miRNA transfection of multipotent and embryonic stem cells with high efficiency and very low toxicity.
Información de suministro: Shipped at room temperatue
Envase: Store at +4 °C