HeLaFect™ Transfection Reagent, OZ Biosciences

Supplier: OZ Biosciences

HF20500 HF21000 HF25000
103258-384EA 224.88 USD
103258-384 103258-386 103258-388
HeLaFect™ Transfection Reagent, OZ Biosciences
Transfection Reagents

HeLaFect is a lipid based transfection reagent specifically developed for HeLa cell transfection.


  • Highly efficient: more than 80% of transfected HeLa cells
  • Ready-to-use: no need for additional buffer
  • Low nucleic acid amount - minimized toxicity
  • High level of nucleic acid compaction
  • Easy and straightforward protocol
  • Compatible with any culture medium: medium change not required


The cationic design of HeLaFect reagent allows high nucleic acid compaction for a high transfection efficiency into HeLa cells.


Información de suministro: Shipped at room temperature


Envase: Store at -20°C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR