CombiMag™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
CM20100 CM20200 CM21000
103258-370EA 105.43 USD
103258-370 103258-372 103258-374
CombiMag™ Transfection Reagent, OZ Biosciences
Transfection Reagents
Based on the Magnetofection technology, CombiMag is the only reagent that enhances the transfection efficiency of any commercial transfection reagents.
  • Ideal to enhance any transfection reagent efficiency without changing your protocol /cells
  • Perfect for primary cells and hard-to-transfect cells
  • Increase any transfection reagent efficiency
  • Non toxic
  • Compatible with and without serum
  • Simple, rapid and easy-to-use
  • For all types of nucleic acids
It can be used with all types of nucleic acids.
Información accesorios: This reagent needs to be used with a magnetic plate.
Información de suministro: Shipped at room temperature
Envase: Store at +4 °C