si3D-Fect™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
STF40250
STF40500
STF41000
103258-232EA
177.37
USD
103258-232
103258-234
103258-236
si3D-Fect™ Transfection Reagent, OZ Biosciences
Transfection Reagents
si3D-Fect is a 3D transfection reagent specifically designed and developed for silencing gene expression in cells cultured on 3D Scaffolds.
- Highly efficient for gene silencing in 3D matrices
- Ideal for any 3D scaffolds (sponges, matrices, inserts)
- Dedicated to short nucleic acid sequences (siRNA, miRNA…)
- Long term gene silencing
- Universal (primary cells and cell lines)
- Serum compatible
Información de suministro: Shipped at room temperature
Envase: Store at +4 °C