3D-FectIN™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
TN30250 TN30500 TN31000
103258-226EA 184.82 USD
103258-226 103258-228 103258-230
3D-FectIN™ Transfection Reagent, OZ Biosciences
Transfection Reagents
3D-FectIN is the newest 3D-Transfection Reagent specifically developed to directly transfect cells cultured in 3D hydrogels.
  • Highly efficient: cell lines and primary cells
  • Specific for 3D Hydrogels
  • Compatible with all types of nucleic acids
  • Completely biodegradable
  • Long term protein expression
  • Serum compatible
3D-FectIN is suitable for all kind of hydrogels and cells.
Información de suministro: Shipped at room temperature
Envase: Store at +4 °C