Guanidinium hydrochloride ≥99.5% for molecular biology
Supplier: Promega Corporation
|
Warning
|
H5381
H5383
PAH5381EA
54.17
USD
PAH5381
PAH5383
Guanidinium hydrochloride ≥99.5% for molecular biology
Guanidinium hydrochloride
Guanidine-HCl, Molecular Grade, is commonly used for the isolation of intact mRNA from tissues or cultured cells.
- Quality tested and certified free of DNase, RNase and protease activity
Form: Fine, colorless or white crystals.
Storage: 15°C to 30°C
Formula:
NH₂C(=NH)NH₂·HCl Melting Pt: 185 °C Density: 1.354 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013026 CAS Number: 50-01-1 EINECS: 200-002-3 Merck Index: 13,04578 |
|
Specification Test Results
Purity | ≥99.5% |
A230 at 6M | ≤0.15 |
A260 at 6M | ≤0.03 |
A280 at 6M | ≤0.02 |
Moisture | ≤0.3% |
Melting Point | 186–188°C |
Lead | ≤5 ppm |
Zinc | ≤1 ppm |
Copper | ≤1ppm |
Iron | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...