Rink Amide Resin, Chem Impex International
Supplier: CHEM IMPEX INTL INC MS
12664-25G
12664-5G
12664-1G
102643-090EA
574.66
USD
102643-090
102643-110
102643-186
Rink Amide Resin, Chem Impex International
Chromatography Sorbents
SPE Sorbents
It is used for the synthesis of the solid phase peptide synthesis of peptide amines utilizing Fmoc-protected amino acids.
- Synonyms: 4-(2',4'-Dimethoxyphenyl-Fmoc-aminomethyl)phenoxy resin
- DVB%: 1%
- Substitution: 0.7-0.9 meq/g (UV spectroscopy after coupling with Fmoc-Leu-OH)
This resin is commonly utilized in combinatorial chemistry to prepare amides, it can be easily cleaved by TFA. This product can be used if a C-terminal amide is desired.