Sodium dodecyl sulphate (SDS), Molecular Biology Grade

Supplier: Promega Corporation

Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

H5113 H5114
PAH5113EA 76.77 USD
PAH5113 PAH5114
Sodium dodecyl sulphate (SDS), Molecular Biology Grade
Sodium dodecyl sulphate (SDS)

A detergent to denature proteins.


  • Quality tested and certified free of DNase and RNase activity


Sodium Dodecyl Sulfate, Molecular Biology Grade (SDS), is a detergent that is known to denature proteins. It is used in denaturing polyacrylamide gel electrophoresis for the determination of protein molecular weight. It is also used in nucleic acid extraction procedures for the disruption of cell walls and dissociation of nucleic acid:protein complexes.

Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR