EDTA disodium salt dihydrate, Molecular Biology Grade, Calbiochem®
Supplier: MilliporeSigma
324503-100GM
324503-1KG
80502-380EA
85.32
USD
80502-380
80502-382
EDTA disodium salt dihydrate, Molecular Biology Grade, Calbiochem®
EDTA disodium salt dihydrate
Formula:
C₁₀H₁₄N₂Na₂O₈·2H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150037 CAS Number: 6381-92-6 |
|
Specification Test Results
Appearance | White crystalline solid |
Contaminants | DNases, proteases, RNases: none detected |
Storage | Store at 15 to 30 °C, Hygroscopic |
Stability | Following reconstitution, store at room temperature (20 °C). Stock solutions are stable for up to 6 months at 20 °C. |
Solubility | H₂O (186 mg/mL or 0.5M) Adjust pH to 8.0 for complete solubilization. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...