Helicobacter sp., Animal IHC Positive Control Slides, Newcomer Supply

Supplier: Newcomer Supply
3274A 3274B
75876-898BX 106.23 USD
75876-898 75876-900
Helicobacter sp., Animal IHC Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Helicobacter sp., Animal Control Slides contain sections of positive staining animal gastrointestinal tract and negative staining human lung.

  • Tissue Type: (+) GI Tract, Animal / (-) Lung
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • IHC slides are cut at 4 µm
  • Standard tissue placement is mid-high on slide

Newcomer Supply Helicobacter sp., Animal Control Slides are for the positive immunohistochemical staining of Helicobacter sp., spiral shaped bacterium strongly associated with inflammation of the stomach and implicated in the development of gastric malignancy, peptic ulcers, and chronic gastritis.

Staining Results:
Helicobacter sp. positive expression will stain golden to dark brown bacteria.
Lung will stain negative.
Nuclei will stain blue.

These positive control slides are intended to be used to verify histological techniques and reagent reactivity. These slides are to be used for the qualitative purpose of determining positive or negative results, and are not intended to be used for any quantitative purpose.

Información para pedidos: All controls are cut in serial sections.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of IHC control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: Allow 2-3 weeks for delivery for large sets. In human tissue unless otherwise indicated.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR