CD56 IHC Positive Control Slides, Newcomer Supply

Supplier: Newcomer Supply
3184A 3184B
75876-866BX 87.8 USD
75876-866 75876-868
CD56 IHC Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
CD56 control slides contain sections of positive staining pancreas and negative staining lung.

  • Tissue type: (+) Pancreas / (-) Lung
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • IHC slides are cut at 4 µm
  • Standard tissue placement is mid-high on slide

Newcomer supply CD56 control slides are for the positive immunohistochemical staining of CD56, expressed in a variety of normal and abnormal tissues

Staining Results:
CD56 positive expression will stain brown membrane staining.
Lung will stain negative.
Nuclei will stain blue.

These positive control slides are intended to be used to verify histological techniques and reagent reactivity. These slides are to be used for the qualitative purpose of determining positive or negative results, and are not intended to be used for any quantitative purpose.

CD56 recognizes two proteins of the neural cell adhesion molecule, the basic molecule expressed on most neuroectodermally derived tissues and neoplasms. It is also expressed on some mesodermally derived tumors.

Información para pedidos: All controls are cut in serial sections.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of IHC control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: Allow 2-3 weeks for delivery for large sets. In human tissue unless otherwise indicated.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR