Colloidal Iron Histo Positive Control Slides, Newcomer Supply

Supplier: Newcomer Supply
4127A 4127B
75876-980BX 75.55 USD
75876-980 75876-982
Colloidal Iron Histo Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Colloidal iron control slides contain a section of positive staining small intestine.

  • Tissue Type: (+) Small Intestine
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • Histo slides are cut at 4 µm, unless otherwise specified
  • Standard tissue placement is mid-high on slide

Newcomer supply colloidal iron control slides are for the positive histochemical staining of acid epithelial mucins (sialomucin, sulfomucin) and stromal (mesenchymal) mucin in tissue sections.

Staining Results:
Acid epithelial mucins will stain blue.
Stromal mucin will stain blue.
Collagen will stain red.
Muscle and cytoplasm will stain yellow.

These positive control slides are intended to be used to verify histological techniques and reagent reactivity. These slides are to be used for the qualitative purpose of determining positive or negative results, and are not intended to be used for any quantitative purpose.

Información para pedidos: All controls are cut in serial sections.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of Histo control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: In human tissue unless otherwise indicated. Histopathology control slides are not quality controlled for IHC methods, as such, we cannot guarantee antigenicity.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR