Blastomyces sp. Histo Positive Control Slides, Newcomer Supply

Supplier: Newcomer Supply
4090A 4090B
75876-964BX 73 USD
75876-964 75876-966
Blastomyces sp. Histo Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Blastomyces sp. control slides contain sections of positive staining lung and negative staining lung.

  • Tissue Type: (+) Lung / (-) Lung
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • Histo slides are cut at 4 µm, unless otherwise specified
  • Standard tissue placement is mid-high on slide

Newcomer Supply Blastomyces sp

Staining Results:
Blastomyces sp. will stain crisp black cell walls & visible internal structures.
Mucin will stain taupe to dark gray.
Background will stain green.
Negative lung will stain negative for fungal organisms.

These positive control slides are intended to be used to verify histological techniques and reagent reactivity. These slides are to be used for the qualitative purpose of determining positive or negative results, and are not intended to be used for any quantitative purpose.

Control slides are for the positive histochemical staining of fungal organisms. The morphology of the organisms is consistent with Blastomyces sp.

Información para pedidos: All controls are cut in serial sections.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of Histo control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: In human tissue unless otherwise indicated. Histopathology control slides are not quality controlled for IHC methods, as such, we cannot guarantee antigenicity.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR