Argentaffin Histo Positive Control Slides, Newcomer Supply

Supplier: Newcomer Supply
4035B 4035A
75876-952BX 561.6 USD
75876-952 75877-298
Argentaffin Histo Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Argentaffin control slides contain a section of positive staining small intestine.

  • Tissue Type: (+) Small intestine
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • Histo slides are cut at 4 µm, unless otherwise specified
  • Standard tissue placement is mid-high on slide

Newcomer supply argentaffin control slides are for the positive histochemical staining of argentaffin substances in tissue sections.

Staining Results:
Melanin and argentaffin granules will stain black.
Nuclei will stain pink-red.

These positive control slides are intended to be used to verify histological techniques and reagent reactivity. These slides are to be used for the qualitative purpose of determining positive or negative results, and are not intended to be used for any quantitative purpose.

Información para pedidos: All controls are cut in serial sections.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of Histo control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: In human tissue unless otherwise indicated. Histopathology control slides are not quality controlled for IHC methods, as such, we cannot guarantee antigenicity.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR