FluoMag™ Transfection Reagents, OZ Biosciences
Supplier: OZ Biosciences
FN10100 FC10100 FP10100 FS10100 FV10100
103258-182EA 191.71 USD
103258-182 103258-184 103258-186 103258-188 103258-190
FluoMag™ Transfection Reagents, OZ Biosciences
Transfection Reagents
OZ Biosciences has created fluorescently-labeled magnetic nanoparticles for Magnetofection applications.
  • Advantages of the Magnetofection technology
  • Easy cell labeling and tracking
  • Non toxic
  • Serum compatible
FluoMag Transfection Reagents are as efficient as their non-labeled counterparts and allow you to visualize the nanoparticles in vitro during your experiments. FluoMag-C corresponding to CombiMag (transfection reagent enhancer), FluoMag-P corresponding to PolyMag (DNA transfection reagent), FluoMag-S corresponding to SilenceMag (siRNA delivery), FluoMag-V corresponding to ViroMag (viral transduction), FluoMag-N corresponding to NeuroMag (neuron transfection).
Información accesorios: This reagent needs to be used with a magnetic plate
Información de suministro: Shipped at room temperature
Envase: Store at +4 °C and protect from the light