In vivo ViroMag™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
In vivo ViroMag Transfection Reagent has been designed for in vivo targeted transduction with any viral vector
- High and increased transduction/infection efficiency
- Magnetically targeted transduction to specific area
- Reduction of virus dose and systemic dissemination
- Work under non permissive conditions (hypothermia, physiological flow conditions)
- Penetration of the vector into tissues
- Minimized toxicity
- Suitable for all types of viruses
Virus/nanoparticles complexes can be easily administrated through various injection routes such as systemic administration (intravenous, intra-artery) or local administration (intratumoral, intracerebroventricular).
Información accesorios: This reagent needs to be used with a magnetic plate.
Información de suministro: Shipped at room temperature
Envase: Store at +4 °C