ViroMag™ R/L Transduction Reagent, OZ Biosciences
Supplier: OZ Biosciences
KC30700 RL41000 RL40100 RL40200
103258-138EA 924.23 USD
103258-138 103258-142 103258-416 103258-418
ViroMag™ R/L Transduction Reagent, OZ Biosciences
Transfection Reagents
ViroMag R/L Transduction Reagent is a magnetic nanoparticles formulation optimized for Retroviruses and Lentiviruses.
  • Increase transduction efficiency
  • Accelerate the transduction process
  • Efficient for hard-to-infect and non-permissive cells
  • Concentrate the viral dose onto the cells
  • Suitable for all type of Retro/Lentivirus
  • Synchronise cells adsorption/infection without modifications of the viruses
Based on the Magnetofection™ technology, this reagent allows concentrating the complete applied dose of Retro/Lentiviral particles onto cells within minutes, inducing a significant improvement of virus infectivity with extremely low vector doses.
Información accesorios: This reagent needs to be used with a magnetic plate.
Información de suministro: Shipped at room temperature
Envase: Store at +4 °C