ViroMag™ Transduction Reagent, OZ Biosciences
Supplier: OZ Biosciences
ViroMag Transduction Reagent is a magnetic nanoparticles formulation dedicated to increase virus infection and transduction capacities; it is suitable for all viruses.
- Increases transduction efficiency
- Accelerates the transduction process
- Efficient for hard-to-infect and non-permissive cells
- Concentrates the viral dose onto the cells
- Suitable for all type of virus
- Synchronise cells adsorption/infection without modifications of the virus
Based on the Magnetofection™ technology, this reagent allows concentrating the complete applied dose of viral particles onto cells within minutes, inducing a significant improvement of virus transduction with extremely low vector doses. Due to its specific properties, ViroMag is ideal to infect non permissive cells.
Información accesorios: This reagent needs to be used with a magnetic plate.
Información de suministro: Shipped at room temperature
Envase: Store at +4 °C