RmesFect™ Transfection Reagent, OZ Biosciences

Supplier: OZ Biosciences

RM20500 RM21000 RM25000
103258-120EA 264.62 USD
103258-120 103258-122 103258-124
RmesFect™ Transfection Reagent, OZ Biosciences
Transfection Reagents

RmesFect Transfection Reagent is based on the TEE-technology specifically designed for mRNA transfection with high efficiency and low toxicity.


  • Highly efficient: achieve mRNA delivery in all cells
  • Ready-to-use: no need for additional buffer
  • Low nucleic acid amount: minimized toxicity
  • Protects mRNA against degradation
  • Compatible with any culture medium
  • Serum compatible: Medium change not required


Información de suministro: Shipped at room temperatue (RmesFect Stem Kit GFP control shipped in dry ice)


Envase: Store at -20°C (RmesFect Stem Kit GFP control store at -80°C)

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR