Propyl benzoate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Benzoic acid propyl ester

H66793-18 H66793-30
AAH66793-18EA 45.04 USD
AAH66793-18 AAH66793-30
Propyl benzoate 99%
Propyl benzoate
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 231°C
Density: 1.026
Flash Pt: 98°(208°F)
Storage Temperature: Ambient
MDL Number: MFCD00009370
CAS Number: 2315-68-6
EINECS: 219-020-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR